Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059975.5 | NP_492376.1 | 5 - 6 | 801 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |