Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026217.4 | NP_001021388.1 | 6 - 7 | 879 | 82 | |
NM_001026218.5 | NP_001021389.1 | 6 - 7 | 885 | 82 | ||
NM_001026219.5 | NP_001021390.1 | 6 - 7 | 830 | 82 | ||
NM_001128984.4 | NP_001122456.2 | 8 - 9 | 1046 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |