Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csnk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060293.6 | NP_492694.1 | 4 - 5 | 594 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |