Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058389.4 | NP_490790.1 | 4 - 5 | 524 | 84 | |
GenBank mRNA | AY008138.1 | - | 4 - 5 | 520 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |