Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058786.7 | NP_491187.2 | 7 - 8 | 1271 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |