Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: him-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047193.5 | NP_001040658.2 | 6 - 7 | 2573 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |