Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spp-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171399.6 | NP_741465.1 | 3 - 4 | 548 | 77 | |
NM_171400.5 | NP_741466.1 | 3 - 4 | 542 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |