Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171645.6 | NP_741740.3 | 13 - 14 | 1843 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Caenorhabditis elegans Probe spans exons |