Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sax-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029819.7 | NP_001024990.1 | 9 - 10 | 3375 | 86 | |
NM_171652.7 | NP_741748.2 | 9 - 10 | 3373 | 86 | ||
GenBank mRNA | AF041053.1 | - | 9 - 10 | 3290 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |