Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373456.2 | NP_001360793.1 | 7 - 8 | 1117 | 64 | |
NM_001373457.2 | NP_001360791.1 | 5 - 6 | 997 | 64 | ||
NM_001373458.3 | NP_001360790.1 | 8 | 1134 | 64 | ||
NM_001373459.2 | NP_001360794.1 | 5 - 6 | 983 | 64 | ||
NM_001373460.3 | NP_001360792.1 | 5 - 6 | 854 | 64 | ||
NM_171758.5 | NP_741880.1 | 6 - 7 | 1017 | 64 | ||
NM_171759.6 | NP_741881.1 | 5 - 6 | 1081 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |