Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060749.2 | NP_493150.2 | 8 - 9 | 1710 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |