Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ero-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377680.2 | NP_001364667.1 | 2 - 3 | 620 | 89 | |
NM_001377681.1 | NP_001364626.1 | 3 - 4 | 515 | 89 | ||
NM_001377682.2 | NP_001364632.1 | 1 - 2 | 96 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |