Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubxn-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313620.3 | NP_001300549.1 | 1 - 2 | 83 | 56 | |
NM_001383772.1 | 6 - 7 | 571 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |