Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: somi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373144.3 | NP_001360520.1 | 2 - 3 | 673 | 61 | |
NM_001373145.2 | NP_001360558.1 | 1 | 576 | 61 | ||
NM_001383439.1 | 3 - 4 | 781 | 61 | |||
NM_073919.6 | NP_506320.3 | 2 - 3 | 671 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |