Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sri-63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062097.1 | NP_494498.1 | 4 - 5 | 808 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |