Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062277.4 | NP_494678.3 | 9 - 10 | 1462 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |