Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267509.3 | NP_001254438.1 | 8 - 9 | 1752 | 85 | |
NM_001377905.1 | NP_001364664.1 | 41 - 42 | 11509 | 85 | ||
NM_064643.10 | NP_497044.3 | 32 - 33 | 9249 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |