Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: feh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373858.2 | NP_001360128.1 | 8 - 9 | 1751 | 85 | |
NM_065177.6 | NP_497578.4 | 10 - 11 | 1915 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |