Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136364.4 | NP_001129836.1 | 9 - 10 | 1194 | 60 | |
NM_065330.7 | NP_497731.1 | 9 - 10 | 1123 | 60 | ||
GenBank mRNA | AF083224.1 | - | 9 - 10 | 1145 | 60 | |
AY204167.1 | - | 8 - 9 | 1124 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |