Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267957.3 | NP_001254886.1 | 21 - 22 | 5345 | 67 | |
NM_001381816.2 | NP_001366699.1 | 20 - 21 | 5279 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |