Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wht-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322779.3 | NP_001309589.1 | 6 | 1023 | 59 | |
NM_001322780.3 | NP_001309591.1 | 4 - 5 | 807 | 59 | ||
NM_001322781.3 | NP_001309590.1 | 4 - 5 | 849 | 59 | ||
NM_065424.3 | NP_497825.2 | 6 - 7 | 1112 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |