Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025942.4 | NP_001021113.1 | 14 - 15 | 2033 | 109 | |
NM_001025943.3 | NP_001021114.1 | 16 - 17 | 2285 | 109 | ||
GenBank mRNA | AF318606.2 | - | 16 - 17 | 2158 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |