Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pfd-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066181.6 | NP_498582.1 | 1 - 2 | 118 | 110 | |
GenBank mRNA | AF303258.1 | - | 1 - 2 | 118 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |