Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066277.5 | NP_498678.3 | 14 - 15 | 4884 | 123 | |
GenBank mRNA | AF245435.1 | - | 14 - 15 | 4861 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |