Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hlh-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066729.9 | NP_499130.2 | 4 - 5 | 586 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |