Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xpo-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307224.3 | NP_001294153.1 | 11 - 12 | 2805 | 114 | |
NM_001380346.1 | NP_001368437.1 | 4 - 5 | 1824 | 114 | ||
NM_068919.8 | NP_501320.2 | 11 - 12 | 2799 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |