Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sax-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038305.3 | NP_001033394.1 | 9 - 10 | 2655 | 80 | |
NM_001038306.5 | NP_001033395.1 | 12 - 13 | 3033 | 80 | ||
NM_001038307.5 | NP_001033396.1 | 12 - 13 | 3043 | 80 | ||
NM_001038308.4 | NP_001033397.1 | 9 - 10 | 2418 | 80 | ||
GenBank mRNA | AB206669.1 | - | 12 - 13 | 2970 | 80 | |
AB206670.1 | - | 9 - 10 | 2418 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |