Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hgrs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068974.8 | NP_501375.2 | 4 - 5 | 355 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |