Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069269.5 | NP_501670.2 | 9 - 10 | 1727 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |