Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pis-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069347.4 | NP_501748.1 | 4 - 5 | 502 | 109 | |
NM_069348.7 | NP_501749.1 | 4 - 5 | 498 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |