Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069396.8 | NP_501797.1 | 4 - 5 | 763 | 77 | |
GenBank mRNA | AB040993.1 | - | 4 - 5 | 717 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |