Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asm-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047531.2 | NP_001040996.1 | 12 - 13 | 1619 | 111 | |
NM_001047532.6 | NP_001040997.1 | 11 - 12 | 1417 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |