Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067575.3 | NP_499976.3 | 2 - 3 | 363 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |