Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306849.3 | NP_001293778.1 | 5 - 6 | 605 | 69 | |
NM_068584.6 | NP_500985.4 | 4 - 5 | 510 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |