Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phy-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069916.6 | NP_502317.1 | 7 - 8 | 1218 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |