Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcap-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307008.3 | NP_001293937.1 | 2 - 3 | 382 | 92 | |
NM_001307009.3 | NP_001293938.1 | 1 - 2 | 112 | 92 | ||
NM_070207.7 | NP_502608.3 | 3 - 4 | 664 | 92 | ||
NM_070208.6 | NP_502609.2 | 2 - 3 | 682 | 92 | ||
GenBank mRNA | DQ143943.1 | - | 3 - 4 | 634 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |