Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snt-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313245.4 | NP_001300174.1 | 2 - 3 | 233 | 116 | |
NM_001313246.3 | NP_001300175.1 | 2 - 3 | 143 | 116 | ||
NM_001313247.3 | NP_001300176.1 | 4 - 5 | 338 | 116 | ||
NM_071008.5 | NP_503409.1 | 4 - 5 | 434 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |