Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srbc-81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071289.2 | NP_503690.2 | 1 - 2 | 636 | 128 |
Gene Symbol: Y19D10A.17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047725.1 | NP_001041190.1 | 1 - 2 | 636 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |