Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ketn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307739.4 | NP_001294668.1 | 24 - 25 | 13597 | 76 | |
NM_001307740.4 | NP_001294669.1 | 23 - 24 | 13474 | 76 | ||
NM_001377591.2 | NP_001364828.1 | 15 - 16 | 11557 | 76 | ||
NM_001377594.2 | NP_001364827.1 | 16 - 17 | 11680 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |