Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330798.2 | NP_001317822.1 | 5 - 6 | 654 | 63 | |
NM_071623.5 | NP_504024.2 | 5 - 6 | 721 | 63 | ||
GenBank mRNA | AY519853.1 | - | 5 - 6 | 657 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |