Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tam-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071934.6 | NP_504335.1 | 4 - 5 | 2165 | 85 | |
GenBank mRNA | AF164625.1 | - | 3 - 4 | 779 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |