Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072979.4 | NP_505380.1 | 7 - 8 | 1731 | 123 |
Set Membership: |
![]() ![]() ![]() ![]() |