Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vab-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129505.5 | NP_001122977.1 | 3 - 4 | 480 | 121 | |
NM_073662.6 | NP_506063.2 | 11 - 12 | 1836 | 121 | ||
NM_073663.6 | NP_506064.1 | 2 - 3 | 608 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |