Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001359794.1 | NP_001346694.1 | 2 - 3 | 520 | 166 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |