Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emb-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269902.2 | NP_001256831.1 | 10 - 11 | 2839 | 96 | |
NM_001277003.3 | NP_001263932.1 | 1 - 2 | 382 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |