Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifta-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076539.6 | NP_508940.4 | 19 - 20 | 2793 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |