Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sul-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373376.2 | NP_001360719.1 | 3 - 4 | 801 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |