Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077020.7 | NP_509421.2 | 10 - 11 | 1559 | 161 |
Set Membership: |
![]() ![]() ![]() ![]() |