Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oct-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029821.6 | NP_001024992.2 | 3 - 4 | 700 | 108 | |
NM_077498.9 | NP_509899.3 | 8 - 9 | 1551 | 108 | ||
GenBank mRNA | AF199013.1 | - | 3 - 4 | 700 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |