Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sym-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078025.7 | NP_510426.1 | 3 - 4 | 540 | 115 | |
GenBank mRNA | AJ242473.1 | - | 3 - 4 | 444 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |